3-(Difluoromethoxy)benzoic acid

Supplier: Matrix Scientific

020858-1G 020858-5G
101810-896EA 110.77 USD
101810-896 101810-898
3-(Difluoromethoxy)benzoic acid
3-(Difluoromethoxy)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00236224
CAS Number: 4837-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR