3-Nitroanthranilic acid ≥97%

Supplier: Matrix Scientific

020615-5G 020615-25G 020615-100G
101810-456EA 15.38 USD
101810-456 101810-458 101810-460
3-Nitroanthranilic acid ≥97%
3-Nitroanthranilic acid

Hazard code(s): Irritant

Formula: C₇H₆N₂O₄
Melting Pt: 207…211 °C
MDL Number: MFCD00024261
CAS Number: 606-18-8

Order Now

Specification Test Results

Melting point 207-211°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR