3-Bromo-2-methylpyridine ≥98%

Supplier: Matrix Scientific

020614-1G 020614-5G 020614-25G
101810-450EA 15.38 USD
101810-450 101810-452 101810-454
3-Bromo-2-methylpyridine ≥98%
3-Bromo-2-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆BrN
Density: 1.495 g/cm³
MDL Number: MFCD00191224
CAS Number: 38749-79-0

Order Now

Specification Test Results

Density 1.495
ND20 1.5605

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR