6-Nitro-o-toluidine ≥99% for synthesis

Supplier: Matrix Scientific
Danger

Synonyms: 6-Nitro-2-methylaniline, 2-Amino-3-nitrotoluene, 2-Methyl-6-nitroaniline

020609-25G 020609-100G
101810-432EA 21.54 USD
101810-432 101810-434
6-Nitro-o-toluidine ≥99% for synthesis
6-Nitro-o-toluidine

Hazard code(s): Toxic, Irritant

Formula: C₇H₈N₂O₂
Boiling Pt: 120 - 124 °C (1 -rr)
Melting Pt: 92 - 94 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007744
CAS Number: 570-24-1
EINECS: 209-329-6
UN: 2660
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 93-96°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR