5-Hydroxy-1-tetralone ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one

020608-1G 020608-5G
101810-428EA 44.62 USD
101810-428 101810-430
5-Hydroxy-1-tetralone ≥98%
5-Hydroxy-1-tetralone

Hazard code(s): Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 209…211 °C
MDL Number: MFCD00001693
CAS Number: 28315-93-7
EINECS: 248-958-0

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR