2-Amino-3-bromopyridine ≥98%

Supplier: Matrix Scientific
Warning

020605-5G 020605-25G
101810-420EA 30.77 USD
101810-420 101810-422
2-Amino-3-bromopyridine ≥98%
2-Amino-3-bromopyridine

Hazard code(s): Irritant

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 63…66 °C
MDL Number: MFCD03095187
CAS Number: 13534-99-1

Order Now

Specification Test Results

Melting point 63-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR