5-Amino-3-fluorobenzonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-fluorobenzonitrile, 3-Cyano-5-fluoroaniline
020601-5G
020601-25G
101810-410EA
41.54
USD
101810-410
101810-412
5-Amino-3-fluorobenzonitrile ≥97%
5-Amino-3-fluorobenzonitrile
Hazard code(s): Irritant
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Boiling Pt: 288 °C Melting Pt: 105…107 °C Density: 1.25 g/cm³ |
MDL Number:
MFCD05864357 CAS Number: 210992-28-2 |
Specification Test Results
Melting point | 105-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...