2-Chloro-4-methylpyrimidine ≥97%
Supplier: Matrix Scientific
020588-5G
020588-25G
101810-378EA
43.08
USD
101810-378
101810-380
2-Chloro-4-methylpyrimidine ≥97%
2-Chloro-4-methylpyrimidine
Hazard code(s): Irritant, Avoid Skin Contact
Formula:
C₅H₅ClN₂ Melting Pt: 45…48 °C |
MDL Number:
MFCD00054434 CAS Number: 13036-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...