5-Bromonicotinonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
5-Bromopyridine-3-carbonitrile, 3-Bromopyridine-5-carbonitrile, 5-Bromo-3-cyanopyridine, 3-Bromo-5-cyanopyridine
020584-1G
020584-5G
020584-25G
101810-358EA
76.92
USD
101810-358
101810-360
101810-362
5-Bromonicotinonitrile ≥98%
5-Bromonicotinonitrile
Hazard code(s): Irritant
Formula:
C₆H₃BrN₂ MW: 183.01 g/mol |
MDL Number:
MFCD00174363 CAS Number: 35590-37-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...