2-Bromo-5-formylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde
020576-1G
020576-5G
101810-336EA
15.38
USD
101810-336
101810-338
2-Bromo-5-formylpyridine ≥97%
2-Bromo-5-formylpyridine
Hazard code(s): Irritant
Formula:
C₆H₄BrNO MW: 186.01 g/mol Boiling Pt: 284 °C Melting Pt: 104…110 °C Density: 1.683 g/cm³ |
MDL Number:
MFCD04115419 CAS Number: 149806-06-4 |
Specification Test Results
Boiling point | 284°C |
Melting point | 104-110°C |
Density | 1.683 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...