2-Chloro-5-formylpyridine ≥97%
Supplier: Matrix Scientific
020575-5G
020575-25G
101810-332EA
30.77
USD
101810-332
101810-334
2-Chloro-5-formylpyridine ≥97%
2-Chloro-5-formylpyridine
Hazard code(s): Irritant
Formula:
C₆H₄ClNO Melting Pt: 77…81 °C |
MDL Number:
MFCD03095223 CAS Number: 23100-12-1 |
Specification Test Results
Melting point | 77-81°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...