2,4-Dibromo-3,5-difluoroaniline ≥98%

Supplier: Matrix Scientific

020553-5G 020553-25G
101810-242EA 30.77 USD
101810-242 101810-244
2,4-Dibromo-3,5-difluoroaniline ≥98%
2,4-Dibromo-3,5-difluoroaniline

Hazard code(s): Irritant

Formula: C₆H₃Br₂F₂N
Melting Pt: 81…83 °C
MDL Number: MFCD06797746
CAS Number: 883549-00-6

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR