2-Amino-5-bromobenzonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-cyanoaniline

020552-25G 020552-100G
101810-238EA 26.15 USD
101810-238 101810-240
2-Amino-5-bromobenzonitrile ≥98%
2-Amino-5-bromobenzonitrile

Hazard code(s): Toxic, Light Sensitive, Refrigerate

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 89…92 °C
MDL Number: MFCD00158946
CAS Number: 39263-32-6
EINECS: 254-387-8

Order Now

Specification Test Results

Melting point 89-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR