2-Amino-6-fluoropyridine ≥98%

Supplier: Matrix Scientific
Warning

020551-5G 020551-25G
101810-234EA 44.62 USD
101810-234 101810-236
2-Amino-6-fluoropyridine ≥98%
2-Amino-6-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD04114200
CAS Number: 1597-32-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR