2-Fluoro-4-methoxybenzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-fluoro-4-cyano anisole, 1-Cyan-2-fluor-4-methoxybenzene

020188-5G 020188-25G
101809-574EA 41.54 USD
101809-574 101809-576
2-Fluoro-4-methoxybenzonitrile ≥97%
2-Fluoro-4-methoxybenzonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD00509374
CAS Number: 94610-82-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR