2-Chloro-6-fluoropyridine ≥98%

Supplier: Matrix Scientific

020183-5G 020183-25G
101809-552EA 15.38 USD
101809-552 101809-554
2-Chloro-6-fluoropyridine ≥98%
2-Chloro-6-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₃ClFN
MDL Number: MFCD00077483
CAS Number: 20885-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR