2-Bromo-6-methoxyphenol ≥97%

Supplier: Matrix Scientific

020027-1G 020027-5G 020027-25G
101809-270EA 69.23 USD
101809-270 101809-272 101809-274
2-Bromo-6-methoxyphenol ≥97%
2-Bromo-6-methoxyphenol

Hazard code(s): Irritant

Formula: C₇H₇BrO₂
MDL Number: MFCD08146628
CAS Number: 28165-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR