1-Boc-piperazine ≥99%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

020026-25G 020026-100G
101809-266EA 26.15 USD
101809-266 101809-268
1-Boc-piperazine ≥99%
1-Boc-piperazine

Hazard code(s): Irritant, Store in Freezer

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Melting Pt: 42…44 °C
MDL Number: MFCD00075265
CAS Number: 57260-71-6

Order Now

Specification Test Results

Melting point 42-44°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR