3-Bromo-5-fluorophenol ≥98%

Supplier: Matrix Scientific

019928-25G 019928-100G
101809-068EA 50.77 USD
101809-068 101809-070
3-Bromo-5-fluorophenol ≥98%
3-Bromo-5-fluorophenol

Hazard code(s): Irritant

Formula: C₆H₄BrFO
MDL Number: MFCD07783710
CAS Number: 433939-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR