5-Bromo-2-iodopyrimidine ≥97%

Supplier: Matrix Scientific

019927-5G 019927-25G
101809-062EA 24.62 USD
101809-062 101809-064
5-Bromo-2-iodopyrimidine ≥97%
5-Bromo-2-iodopyrimidine

Hazard code(s): Irritant, Light Sensitive

Formula: C₄H₂BrIN₂
MW: 284.88 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD01318111
CAS Number: 183438-24-6

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR