6-Aminoindole ≥98% for synthesis

Supplier: Matrix Scientific

018336-1G 018336-5G
101805-962EA 44.62 USD
101805-962 102832-872
6-Aminoindole ≥98% for synthesis
6-Aminoindole

Hazard code(s): Irritant-Harmful, Store in Freezer

Formula: C₈H₈N₂
MW: 132.16
Boiling Pt: 161…166 °C (2mm)
Melting Pt: 65…66 °C
Storage Temperature: Freezer
MDL Number: MFCD02093959
CAS Number: 5318-27-4

Order Now

Specification Test Results

Boiling point 161-166°C/2
Melting point 65-66°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR