4-Hydroxyindole ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol

018324-5G 018324-100G
101805-920EA 115.38 USD
101805-920 102663-608
4-Hydroxyindole ≥98%
4-Hydroxyindole

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 100 °C (0.01 mmHg)
Melting Pt: 97…99 °C
MDL Number: MFCD00005667
CAS Number: 2380-94-1
EINECS: 219-177-2

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR