4-Methoxyindole ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Methoxy-1H-indole, Indol-4-yl methyl ether
018277-5G
018277-25G
101805-798EA
269.23
USD
101805-798
102832-864
4-Methoxyindole ≥98%
4-Methoxyindole
Hazard code(s): Store in Freezer, Light Sensitive
Formula:
C₉H₉NO MW: 147.18 g/mol Boiling Pt: 181…183 °C (24mm) Melting Pt: 69…71 °C |
MDL Number:
MFCD00009737 CAS Number: 4837-90-5 |
Specification Test Results
Boiling point | 181-183°C/2 |
Melting point | 69-71°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...