2-Bromo-6-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

018273-5G 018273-100G
101805-778EA 15.38 USD
101805-778 101805-782
2-Bromo-6-fluorobenzonitrile ≥98%
2-Bromo-6-fluorobenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₇H₃BrFN
Melting Pt: 58…62 °C
MDL Number: MFCD06797753
CAS Number: 79544-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR