2-Chloro-5-iodopyridine ≥97%

Supplier: Matrix Scientific
Warning

018206-1G 018206-25G
101805-638EA 15.38 USD
101805-638 101805-642
2-Chloro-5-iodopyridine ≥97%
2-Chloro-5-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD01863635
CAS Number: 69045-79-0

Order Now

Specification Test Results

Melting point 95.8-95.9°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR