3-Methylbenzo[b]thiophene-2-carboxaldehyde ≥97%
Supplier: Matrix Scientific
016036-1G
016036-5G
101803-962EA
69.23
USD
101803-962
101803-964
3-Methylbenzo[b]thiophene-2-carboxaldehyde ≥97%
3-Methylbenzo[b]thiophene-2-carboxaldehyde
Hazard code(s): Irritant, Air Sensitive, Refrigerate
Formula:
C₁₀H₈OS MW: 176.24 g/mol Melting Pt: 88…90 °C |
MDL Number:
MFCD00052307 CAS Number: 22053-74-3 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...