3-Amino-2-methoxypyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Methoxypyridin-3-amine, 2-Methoxy-3-pyridylamine

016034-1G 016034-5G
101803-952EA 15.38 USD
101803-952 101803-954
3-Amino-2-methoxypyridine ≥98%
3-Amino-2-methoxypyridine

Hazard code(s): Irritant

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD00833386
CAS Number: 20265-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR