3-Bromoisonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromopyridine-4-carboxylic acid, 3-Bromo-4-pyridinecarboxylic acid

016033-5G 016033-25G 016033-100G
101803-946EA 16.92 USD
101803-946 101803-948 101803-950
3-Bromoisonicotinic acid ≥97%
3-Bromoisonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 236…238.5 °C
MDL Number: MFCD00040944
CAS Number: 13959-02-9

Order Now

Specification Test Results

Melting point 236-238.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR