3-Bromo-5-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

016029-5G 016029-25G
101803-928EA 75.38 USD
101803-928 101803-930
3-Bromo-5-fluorobenzonitrile ≥98%
3-Bromo-5-fluorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Melting Pt: 43…46 °C
MDL Number: MFCD04038227
CAS Number: 179898-34-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 43-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR