2-Aminonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

016028-25G 016028-100G
101803-924EA 64.62 USD
101803-924 101803-926
2-Aminonicotinic acid ≥98%
2-Aminonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 296…298 °C
Density: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS Number: 5345-47-1
EINECS: 226-296-3

Order Now

Specification Test Results

Melting point 296-298°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR