ω,ω-Difluoroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluoroacetophenone, 2,2-Difluoro-1-phenylethanone, α,α-Difluoroacetophenone

012901-1G 012901-5G
101803-586EA 150.77 USD
101803-586 101803-588
ω,ω-Difluoroacetophenone ≥95%
ω,ω-Difluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 53…54 °C (12mm)
MDL Number: MFCD06246879
CAS Number: 395-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR