2-Aminonicotinonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine

012821-5G 012821-25G
101803-456EA 52.31 USD
101803-456 101803-458
2-Aminonicotinonitrile ≥98%
2-Aminonicotinonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 134 °C
MDL Number: MFCD00128867
CAS Number: 24517-64-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR