Ethyl-5-bromonicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-bromonicotinate

012820-25G 012820-100G
101803-452EA 29.23 USD
101803-452 101803-454
Ethyl-5-bromonicotinate ≥97%
Ethyl-5-bromonicotinate

Hazard code(s): Irritant

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Boiling Pt: 86…92 °C (0.5mm)
Melting Pt: 40…41 °C
MDL Number: MFCD00040366
CAS Number: 20986-40-7

Order Now

Specification Test Results

Boiling point 86-92°C/0.5
Melting point 40-41°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR