2-Bromo-3,5-dichloropyridine ≥98%

Supplier: Matrix Scientific

012675-5G 012675-25G 012675-100G
101803-182EA 15.38 USD
101803-182 101803-184 102832-848
2-Bromo-3,5-dichloropyridine ≥98%
2-Bromo-3,5-dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₂BrCl₂N
Melting Pt: 40…42 °C
MDL Number: MFCD00233990
CAS Number: 14482-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR