(±)-Pipecolinic acid ≥99%

Supplier: Matrix Scientific

Synonyms: (±)-2-Piperidinecarboxylic acid, DL-Pipecolinic acid, H-DL-Homopro-OH, Pipecolinic Acid, Piperidine-2-carboxylic acid, Pipecolinic acid

012673-25G 012673-100G
101803-174EA 20 USD
101803-174 101803-176
(±)-Pipecolinic acid ≥99%
(±)-Pipecolinic acid

Hazard code(s): Irritant

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: ∼280 °C (dec.)
MDL Number: MFCD00064347
CAS Number: 535-75-1
EINECS: 208-616-3

Order Now

Specification Test Results

Melting point ca 280°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR