6-Chloronicotinonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-cyanopyridine, 6-Chloro-3-pyridinecarbonitrile, 2-Chloropyridine-5-carbonitrile
012671-25G
012671-100G
101803-170EA
75.38
USD
101803-170
101803-172
6-Chloronicotinonitrile ≥98%
6-Chloronicotinonitrile
Hazard code(s): Irritant
Formula:
C₆H₃ClN₂ MW: 138.56 g/mol Boiling Pt: 105…107 °C (1mm) Melting Pt: 117…119 °C |
MDL Number:
MFCD00084941 CAS Number: 33252-28-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Boiling point | 105-107°C/1 |
Melting point | 117-119°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...