Ethyl-2-iodobenzoate ≥98%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-iodobenzoate, 2-Iodobenzoic acid ethyl ester
012667-25G
012667-100G
101803-154EA
36.92
USD
101803-154
101803-156
Ethyl-2-iodobenzoate ≥98%
Ethyl-2-iodobenzoate
Hazard code(s): Irritant, Light Sensitive
Formula:
C₉H₉IO₂ MW: 276.07 g/mol Boiling Pt: 163…165 °C (23mm) |
MDL Number:
MFCD00051796 CAS Number: 1829-28-3 |
Specification Test Results
Boiling point | 163-165°C/2 |
ND20 | 1.584 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...