Ethyl-3-iodobenzoate ≥99%

Supplier: Matrix Scientific

Synonyms: 3-Iodobenzoic acid ethyl ester

012666-5G 012666-25G 012666-100G
101803-148EA 15.38 USD
101803-148 101803-150 101803-152
Ethyl-3-iodobenzoate ≥99%
Ethyl-3-iodobenzoate

Hazard code(s): Irritant, Light Sensitive

Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 165 °C (1013 hPa)
Density: 1.66 g/cm³ (20 °C)
MDL Number: MFCD00044769
CAS Number: 58313-23-8

Order Now

Specification Test Results

Boiling point 160-161°C/3
Density 1.66
ND20 1.582

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR