2-Acetyl-3-methylthiophene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(3-Methyl-2-thienyl)-1-ethanone, 1-(3-Methylthiophen-2-yl)ethanone

012657-5G 012657-25G
101803-122EA 73.85 USD
101803-122 101803-124
2-Acetyl-3-methylthiophene ≥98%
2-Acetyl-3-methylthiophene

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 68…72 °C (3mm)
Density: 1.1331 g/cm³ (22.5 °C)
MDL Number: MFCD00005443
CAS Number: 13679-72-6
EINECS: 237-179-1

Order Now

Specification Test Results

Boiling point 68-72°C/3mm
Density 1.126
ND20 1.562

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR