2-Amino-6-chloropurine ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine
012654-25G
012654-100G
101803-114EA
72.31
USD
101803-114
101803-116
2-Amino-6-chloropurine ≥97%
2-Amino-6-chloropurine
Hazard code(s): Irritant, Store in Freezer
Formula:
C₅H₄ClN₅ MW: 169.57 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00075252 CAS Number: 10310-21-1 EINECS: 233-686-7 |
Specification Test Results
Melting point | >300°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...