2-Amino-3-hydroxypyridin ≥98%

Supplier: Matrix Scientific
Danger

Synonyms: 2-Aminopyridin-3-ol, 2-Amino-3-hydroxypyridine

012652-100G 012652-500G
101803-104EA 60 USD
101803-104 101803-106
2-Amino-3-hydroxypyridin ≥98%
2-Amino-3-hydroxypyridin

Hazard code(s): Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006317
CAS Number: 16867-03-1
EINECS: 240-886-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR