3-Methoxy-2-nitropyridine ≥98%

Supplier: Matrix Scientific

012650-25G 012650-100G
101803-100EA 43.08 USD
101803-100 101803-102
3-Methoxy-2-nitropyridine ≥98%
3-Methoxy-2-nitropyridine

Hazard code(s): Irritant

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 73…76 °C
MDL Number: MFCD00222276
CAS Number: 20265-37-6

Order Now

Specification Test Results

Melting point 73-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR