Pyridine-2,6-dicarboxylic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA

012649-25G 012649-100G
101803-096EA 53.85 USD
101803-096 101803-098
Pyridine-2,6-dicarboxylic acid ≥98%
Pyridine-2,6-dicarboxylic acid

Hazard code(s): Irritant

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: ∼249 °C (dec.)
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3

Order Now

Specification Test Results

Melting point ca 249°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR