(4-Aminophenyl)acetic acid ≥98%

Supplier: Matrix Scientific

012647-25G 012647-100G
101803-088EA 35.38 USD
101803-088 101803-090
(4-Aminophenyl)acetic acid ≥98%
(4-Aminophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄(NH₂))CH₂COOH
MW: 151.16 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 201 °C (dec.)
MDL Number: MFCD00007916
CAS Number: 1197-55-3
EINECS: 214-828-7
Merck Index: 13,00462

Order Now

Specification Test Results

Melting point 201°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR