Pyridine-2,3-dicarboxylic acid ≥99%

Supplier: Matrix Scientific

Synonyms: Quinolinic acid

012643-25G 012643-100G
101803-074EA 24.62 USD
101803-074 101803-076
Pyridine-2,3-dicarboxylic acid ≥99%
Pyridine-2,3-dicarboxylic acid

Hazard code(s): Irritant

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: ∼188 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006295
CAS Number: 89-00-9
EINECS: 201-874-8
Merck Index: 12,08258

Order Now

Specification Test Results

Melting point ca 188°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR