2-Bromo-6-fluoroaniline ≥98%

Supplier: Matrix Scientific
Warning

012633-5G 012633-25G 012633-100G
101803-034EA 29.23 USD
101803-034 101803-036 101803-038
2-Bromo-6-fluoroaniline ≥98%
2-Bromo-6-fluoroaniline

Hazard code(s): Toxic

Formula: C₆H₅BrFN
Density: 1.674 g/cm³
MDL Number: MFCD01310982
CAS Number: 65896-11-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR