1,3-Difluoro-5-iodobenzene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3,5-Difluoroiodobenzene

012630-5G 012630-25G 012630-100G
101803-022EA 33.85 USD
101803-022 101803-024 101803-026
1,3-Difluoro-5-iodobenzene ≥98%
1,3-Difluoro-5-iodobenzene

Hazard code(s): Irritant

Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 173…174 °C
MDL Number: MFCD03094155
CAS Number: 2265-91-0

Order Now

Specification Test Results

Boiling point 173-174°C
ND20 1.554

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR