1,4-Difluoro-2-iodobenzene ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2,5-Difluoroiodobenzene

012628-5G 012628-25G 012628-100G
101803-012EA 15.38 USD
101803-012 101803-014 101803-016
1,4-Difluoro-2-iodobenzene ≥98% for synthesis
1,4-Difluoro-2-iodobenzene

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₃F₂I
MW: 239.99
Boiling Pt: 178…179 °C
Density: 1.998 g/cm³
MDL Number: MFCD03094151
CAS Number: 2265-92-1

Order Now

Specification Test Results

Boiling point 178-179°C
Density 1.998
ND20 1.56


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR