2-Fluoro-6-nitrobenzoic acid

Supplier: Matrix Scientific

012622-5G 012622-25G
101802-998EA 29.23 USD
101802-998 101803-000
2-Fluoro-6-nitrobenzoic acid
2-Fluoro-6-nitrobenzoic acid
Formula: C₇H₄FNO₄
MDL Number: MFCD01862079
CAS Number: 385-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR