(±)-2-Fluoromandelic acid ≥98% for synthesis

Supplier: Matrix Scientific

012619-5G 012619-25G
101802-986EA 24.62 USD
101802-986 101802-988
(±)-2-Fluoromandelic acid ≥98% for synthesis
(±)-2-Fluoromandelic acid

Hazard code(s): Irritant

Formula: C₈H₇FO₃
Melting Pt: 114…116 °C
MDL Number: MFCD03095382
CAS Number: 389-31-1

Order Now

Specification Test Results

Melting point 114-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR